missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Sodium 1-octanesulfonate, 99%, Thermo Scientific Chemicals
28.06 GBP valid until 2024-03-29
Use promo code "21615" to get your promotional price.
Use promo code "21615" to get your promotional price.
Click to view available options
Quantity:
5 g
25 g
100 g
Description
Sodium-1-octanesulfonate is used as a reagent for HPLC analysis of organic compounds, and more specifically, peptides and proteins. It is also extensively used as an ion pairing agent. Sodium-1-octanesulfonate is found to interact with human serum albumin. It is also used as an ion complexation agent for N-nitrosodiethanolamine.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
Sodium-1-octanesulfonate is used as a reagent for HPLC analysis of organic compounds, and more specifically, peptides and proteins. It is also extensively used as an ion pairing agent. Sodium-1-octanesulfonate is found to interact with human serum albumin. It is also used as an ion complexation agent for N-nitrosodiethanolamine.
Solubility
Soluble in water producing a clear and colorless solution
Notes
Sodium D-gluconate is stable under normal temperatures.
Sodium-1-octanesulfonate is used as a reagent for HPLC analysis of organic compounds, and more specifically, peptides and proteins. It is also extensively used as an ion pairing agent. Sodium-1-octanesulfonate is found to interact with human serum albumin. It is also used as an ion complexation agent for N-nitrosodiethanolamine.
Solubility
Soluble in water producing a clear and colorless solution
Notes
Sodium D-gluconate is stable under normal temperatures.
Specifications
Melting Point | >300°C |
Assay Percent Range | 99% |
Quantity | 5 g |
Beilstein | 3574241 |
Solubility Information | Soluble in water producing a clear and colorless solution |
Formula Weight | 216.28 |
Chemical Name or Material | Sodium 1-octanesulfonate |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'