missing translation for 'onlineSavingsMsg'
Learn More

Sodium 1-heptanesulfonate monohydrate, 99%, Thermo Scientific Chemicals

Catalog Number p-7049583
Click to view available options
Quantity:
10 g
50 g
207300-90-1
C7H17NaO4S
220.259
MFCD00149550
XWZCREJRXRKIRQ-UHFFFAOYSA-M
sodium heptane-1-sulfonate hydrate, 1-heptanesulfonic acid sodium salt monohydrate, sodium 1-heptanesulfonate monohydrate, sodium 1-heptanesulfonate hydrate, sodium1-heptanesulfonatemonohydrate, potassium heptane-1-sulfonate hydrate, sodium heptane-1-sulfonate-water 1/1/1, sodium 1-heptanesulfonate monohydrate t, sodium 1-heptanesulfonate monohydrate, for hplc
23687711
sodium;heptane-1-sulfonate;hydrate
CCCCCCCS(=O)(=O)[O-].O.[Na+]
207300-90-1
C7H17NaO4S
220.259
MFCD00149550
XWZCREJRXRKIRQ-UHFFFAOYSA-M
sodium heptane-1-sulfonate hydrate, 1-heptanesulfonic acid sodium salt monohydrate, sodium 1-heptanesulfonate monohydrate, sodium 1-heptanesulfonate hydrate, sodium1-heptanesulfonatemonohydrate, potassium heptane-1-sulfonate hydrate, sodium heptane-1-sulfonate-water 1/1/1, sodium 1-heptanesulfonate monohydrate t, sodium 1-heptanesulfonate monohydrate, for hplc
23687711
sodium;heptane-1-sulfonate;hydrate
CCCCCCCS(=O)(=O)[O-].O.[Na+]

Sodium 1-heptanesulfonate monohydrate is an ion pairing reagent used in the HPLC analysis of proteins and peptides. It is utilized in high performance capillary electrophoresis analysis of peptides.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Melting Point >300°C
Color White
Linear Formula CH3(CH2)6SO3Na·H2O
Quantity 10 g
Beilstein 3731762
Solubility Information Slightly soluble in water.
Formula Weight 220.26 (202.24 Anhydrous)
Percent Purity 99%
Physical Form Crystalline
Chemical Name or Material Sodium 1-heptanesulfonate monohydrate

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'