missing translation for 'onlineSavingsMsg'
Learn More

Soda lime, indicating, ACS, Thermo Scientific Chemicals

Catalog Number p-7059489
Click to view available options
Quantity:
500 g
2.5 kg
8006-28-8
CaHNaO2
96.074
MFCD00134094
HUAUNKAZQWMVFY-UHFFFAOYSA-M
66545795
sodium;oxocalcium;hydroxide
[OH-].O=[Ca].[Na+]
8006-28-8
CaHNaO2
96.074
MFCD00134094
HUAUNKAZQWMVFY-UHFFFAOYSA-M
66545795
sodium;oxocalcium;hydroxide
[OH-].O=[Ca].[Na+]

Soda lime's ability is to absorb carbon dioxide, makes it valuable in medical and surgical procedures. It is also used as a powerful drying agent, or desiccant, in commercial and industrial usage. It is also useful for the construction of gas masks. It plays an essential role for the measurement of soil carbon dioxide efflux upon absorption. It is also useful for direct measurement of dissolved carbon dioxide.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY
Physical Form 4-8 Mesh
Material Soda Lime
Quantity 500 g
Mesh Size 4 to 8 Mesh
UN Number UN1907
Merck Index 14,8569
Odor Odorless
Solubility Information Soluble in water.
Sensitivity Air Sensitive
Grade ACS
Chemical Name or Material Soda lime, Indicating
Show More Show Less

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'