Learn More
Hirschmann Instruments™ Clear Glass Graduated Pipettes with Amber Stain Graduation
Provides volumetric accuracy for your pipetting needs. Hirschmann Instruments™ Clear Glass Graduated Pipettes with Amber Stain Graduation come with a cotton plug end as standard.
Brand: Hirschmann Instruments™ 1000141
Code : A0
Additional Details : Weight : 0.25000kg
Description
- Short line
- Acc. to DIN EN ISO 835
- Graduated to tip, zero on top, type 3
Specifications
B | |
Amber | |
360 ± 5mm | |
Graduated Pipette | |
Soda Lime Glass |
Yes | |
Amber graduations permanently with silver diffusion color | |
12Pack | |
2mL |