missing translation for 'onlineSavingsMsg'
Learn More

Hirschmann Instruments™ Clear Glass Graduated Pipettes with Amber Stain Graduation

Provides volumetric accuracy for your pipetting needs. Hirschmann Instruments™ Clear Glass Graduated Pipettes with Amber Stain Graduation come with a cotton plug end as standard.

Brand:  Hirschmann Instruments™ 1000129

Code : A0

Additional Details : Weight : 0.25000kg

 View more versions of this product

Product Code. 11726622

  • £45.50 / Pack of 12


Estimated Shipment
25-06-2018
Add to basket

Description

Description

  • Short line
  • Acc. to DIN EN ISO 835
  • Graduated to tip, zero on top, type 3
Specifications

Specifications

B
Amber
360 ± 5mm
Graduated Pipette
Soda Lime Glass
Yes
Amber graduations permanently with silver diffusion color
12Pack
0.5mL
Videos