missing translation for 'onlineSavingsMsg'
Learn More

Poulten Graf™ Soda Lime Glass Graduated Pipet

Brand:  Poulten Graf™ R388/F/5

Code : 01

Additional Details : Weight : 0.11844kg

Product Code. 12314438

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »


Description

Description

Specifications

Specifications

Soda Lime Glass
5/Pk.
175 mm
Yes
5 mL
Graduated Pipette
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers