missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SNTG1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA567236
Additional Details : Weight : 0.01000kg
Description
Immunogen sequence: LLHSRFSQYVP GTDLSRQNAF QVIAVDGVCT GIIQCLSAED CVDWLQAIAT NISNLTKHNI KKINRNFPVN QQIVY Highest antigen sequence identity to the following orthologs - mouse 92%, rat 93%.
SNTG1 Polyclonal antibody specifically detects SNTG1 in Human samples. It is validated for Immunohistochemistry.Specifications
SNTG1 | |
Polyclonal | |
Unconjugated | |
SNTG1 | |
4933426D16Rik; G1SYN; Gamma-1-syntrophin; gamma1-syntrophin; RGD1560290; Sntg1; SYN4; syntrophin 4; syntrophin gamma 1; syntrophin, gamma 1; syntrophin-4; zmp:0000001117 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
54212 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.05 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NSN8 | |
SNTG1 | |
Recombinant Human SNTG1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
For Research Use Only