missing translation for 'onlineSavingsMsg'
Learn More

SNOG, Tocris Bioscience™

NO carrier. Breaks down to release NO

Brand:  Tocris (Bio-Techne) 060350

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13447803

  • £257.25 / 50mg

Estimated Shipment date
from Supplier 27-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

A carrier of NO, relaxing smooth muscle and inhibiting platelet aggregation.
Specifications

Chemical Identifiers

57564-91-7
336.319
s-nitrosoglutathione, gsno, snog, nitrosoglutathione, glutathione thionitrite, n-n-l-gamma-glutamyl-s-nitroso-l-cysteinyl glycine, s-nitroso-l-glutathione, ccris 2095, s-2-amino-5-r-1-carboxymethyl amino-3-nitrosothio-1-oxopropan-2-yl amino-5-oxopentanoic acid, unii-78rri89zto
CHEBI:50091
C(CC(=O)NC(CSN=O)C(=O)NCC(=O)O)C(C(=O)O)N
C10H16N4O7S
HYHSBSXUHZOYLX-WDSKDSINSA-N
104858
(2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-3-nitrososulfanyl-1-oxopropan-2-yl]amino]-5-oxopentanoic acid

Specifications

50mg
57564-91-7
s-nitrosoglutathione, gsno, snog, nitrosoglutathione, glutathione thionitrite, n-n-l-gamma-glutamyl-s-nitroso-l-cysteinyl glycine, s-nitroso-l-glutathione, ccris 2095, s-2-amino-5-r-1-carboxymethyl amino-3-nitrosothio-1-oxopropan-2-yl amino-5-oxopentanoic acid, unii-78rri89zto
C(CC(=O)NC(CSN=O)C(=O)NCC(=O)O)C(C(=O)O)N
336.319
CHEBI:50091
>98%
SNOG
C10H16N4O7S
HYHSBSXUHZOYLX-WDSKDSINSA-N
(2S)-2-amino-5-[[(2R)-1-(carboxymethylamino)-3-nitrososulfanyl-1-oxopropan-2-yl]amino]-5-oxopentanoic acid
104858
336.32
Videos
Documents