Learn More
SNIP Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA564135
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: EGVWPPPNNL LSQSPKKVTA ETDFNKSVDF EMPPPSPPLN LHELSGPAEG ASLTPKGGNP TKGLDTPGKR SVDKAVSVEA AERDWE Highest antigen sequence identity to the following orthologs: Mouse - 81%, Rat - 85%.
SNIP Polyclonal antibody specifically detects SNIP in Human samples. It is validated for Immunocytochemistry, Immunohistochemistry (Paraffin).Specifications
SNIP | |
Polyclonal | |
Unconjugated | |
SRCIN1 | |
KIAA1684; mKIAA1684; p130Cas-associated protein; P140; p140Cap; SNAP-25-interacting protein; SNAP25-interacting protein; SNIP; SRC kinase signaling inhibitor 1; SRCIN1 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
80725 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry, Immunohistochemistry (Paraffin) | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9C0H9 | |
SRCIN1 | |
Recombinant protein corresponding to Human SNIP | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only