missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ Human SNF1LK2, GST Tag Recombinant Protein
SNF1LK2 (QIK) belongs to the AMP-activated protein kinase family.
£429.00 - £6900.00
Specifications
Accession Number | NP_053006.1 |
---|---|
Gene Alias | SNF1LK2, QIK |
KinaseFamily | CAMKL Kinase Family |
Species | Human |
Recombinant | Recombinant |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
10064044
|
Invitrogen™
PV4792 |
10μg |
£429.00
Each |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15428673
|
Invitrogen™
PR8353A |
100μg |
£1602.00
100µg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15488753
|
Invitrogen™
PV4793 |
1mg |
£6900.00
1mg |
Estimated Shipment: 02-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
Description
SNF1LK2 (QIK) is a serine/threonine kinase and is a salt-inducible kinase.
Specifications
NP_053006.1 | |
CAMKL Kinase Family | |
Recombinant | |
Baculovirus |
SNF1LK2, QIK | |
Human | |
GST Tag |