missing translation for 'onlineSavingsMsg'
Learn More

SNC 80, Tocris Bioscience™

Highly selective non-peptide δ agonist

Brand:  Tocris (Bio-Techne) 0764/10

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13457973

  • £183.48 / 10mg


Estimated Shipment
06-03-2019
Add to basket

Description

Description

A highly selective and potent non-peptide δ-opioid agonist, 2000-fold selective over μ-opioid receptors.
Specifications

Chemical Identifiers

156727-74-1
449.639
snc 80, snc-80, chembl13470, snc80, 4-alpha-4-allyl-2,5-dimethyl-1-piperazinyl-3-methoxybenzyl-n,n-diethylbenzamide, dsstox_cid_25662, dsstox_rid_81039, dsstox_gsid_45662, 4-2,5-dimethyl-4-2-propenyl-1-piperazinyl 3-methoxyphenyl methyl-n,n-diethylbenzamide 2s-1 s* ,2alpha,5beta, +-4-alphar-alpha-2s,5r-4-allyl-2,5-dimethyl-1-piperazinyl-3-methoxybenzyl-n,n-diethylbenzamide
4-[(R)-[(2S,5R)-2,5-dimethyl-4-prop-2-enylpiperazin-1-yl]-(3-methoxyphenyl)methyl]-N,N-diethylbenzamide
C28H39N3O2
KQWVAUSXZDRQPZ-UMTXDNHDSA-N
123924
CCN(CC)C(=O)C1=CC=C(C=C1)C(C2=CC(=CC=C2)OC)N3CC(N(CC3C)CC=C)C

Specifications

SNC 80
156727-74-1
snc 80, snc-80, chembl13470, snc80, 4-alpha-4-allyl-2,5-dimethyl-1-piperazinyl-3-methoxybenzyl-n,n-diethylbenzamide, dsstox_cid_25662, dsstox_rid_81039, dsstox_gsid_45662, 4-2,5-dimethyl-4-2-propenyl-1-piperazinyl 3-methoxyphenyl methyl-n,n-diethylbenzamide 2s-1 s* ,2alpha,5beta, +-4-alphar-alpha-2s,5r-4-allyl-2,5-dimethyl-1-piperazinyl-3-methoxybenzyl-n,n-diethylbenzamide
CCN(CC)C(=O)C1=CC=C(C=C1)C(C2=CC(=CC=C2)OC)N3CC(N(CC3C)CC=C)C
449.639
449.64
10mg
C28H39N3O2
KQWVAUSXZDRQPZ-UMTXDNHDSA-N
4-[(R)-[(2S,5R)-2,5-dimethyl-4-prop-2-enylpiperazin-1-yl]-(3-methoxyphenyl)methyl]-N,N-diethylbenzamide
123924
>99%
Videos