missing translation for 'onlineSavingsMsg'
Learn More

SNC 162, Tocris Bioscience™

Potent, selective non-peptide δ agonist

Brand:  Tocris (Bio-Techne) 152910

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13449613

  • £227.85 / 10mg

Estimated Shipment date
from Supplier 21-11-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent and selective non-peptide δ-opioid receptor agonist (Ki = 0.63 nM). Displays > 8000-fold selectivity over μ-opioid receptors and is centrally active following systemic administration in vivo.
Specifications

Chemical Identifiers

178803-51-5
419.613
tocris-1529, 4-s-2s,5r-2,5-dimethyl-4-2-propenyl-1-piperazinyl phenylmethyl-n,n-diethylbenzamide, n,n-diethyl-4-s-alpha-2s-2beta,5alpha-dimethyl-4-allylpiperazino benzyl benzamide, 4-alpha-s-alpha-2s,5r-4-allyl-2,5-dimethyl-1-piperazinyl benzyl-n,n-diethyl benzamide, 4-alpha-s-alpha-2s,5r-4-allyl-2,5-dimethyl-1-piperazinyl benzyl-n,n-diethylbenzamide, 4-s-2s,5r-2,5-dimethyl-4-prop-2-enylpiperazin-1-yl-phenylmethyl-n,n-diethylbenzamide
4-[(S)-[(2S,5R)-2,5-dimethyl-4-prop-2-enylpiperazin-1-yl]-phenylmethyl]-N,N-diethylbenzamide
C27H37N3O
WGIDFDFAOQVAHY-UFPGJGBJSA-N
6604878
CCN(CC)C(=O)C1=CC=C(C=C1)C(C2=CC=CC=C2)N3CC(N(CC3C)CC=C)C

Specifications

10mg
178803-51-5
tocris-1529, 4-s-2s,5r-2,5-dimethyl-4-2-propenyl-1-piperazinyl phenylmethyl-n,n-diethylbenzamide, n,n-diethyl-4-s-alpha-2s-2beta,5alpha-dimethyl-4-allylpiperazino benzyl benzamide, 4-alpha-s-alpha-2s,5r-4-allyl-2,5-dimethyl-1-piperazinyl benzyl-n,n-diethyl benzamide, 4-alpha-s-alpha-2s,5r-4-allyl-2,5-dimethyl-1-piperazinyl benzyl-n,n-diethylbenzamide, 4-s-2s,5r-2,5-dimethyl-4-prop-2-enylpiperazin-1-yl-phenylmethyl-n,n-diethylbenzamide
CCN(CC)C(=O)C1=CC=C(C=C1)C(C2=CC=CC=C2)N3CC(N(CC3C)CC=C)C
419.613
419.61
SNC 162
C27H37N3O
WGIDFDFAOQVAHY-UFPGJGBJSA-N
4-[(S)-[(2S,5R)-2,5-dimethyl-4-prop-2-enylpiperazin-1-yl]-phenylmethyl]-N,N-diethylbenzamide
6604878
>99%
Videos
Documents