missing translation for 'onlineSavingsMsg'
Learn More

SNAP, Tocris Bioscience™

A stable analog of endogenous S-nitroso compounds

Brand:  Tocris (Bio-Techne) 0598/50

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13487793

  • £253.00 / 50mg


Estimated Shipment
08-03-2018
Add to basket

Description

Description

A stable analog of endogenous S-nitroso compounds. A source of NO in vivo which unlike organic O-nitrates does not induce tolerance. Decomposes slowly in solution with a t½ of 37h.
Specifications

Chemical Identifiers

SNAP
C7H12N2O4S
220.24
(S)-Nitroso-N-acetylpenicillamine

Specifications

SNAP
>98%
C7H12N2O4S
(S)-Nitroso-N-acetylpenicillamine
79032-48-7
50mg
220.24
Videos