Learn More
Snail1 Monoclonal Antibody (20C8), eBioscience™, Invitrogen™
Mouse Monoclonal Antibody
Brand: Affymetrix eBioscience 14-9859-82
Code : NEW
Additional Details : Weight : 0.09500kg
Description
Description: The monoclonal antibody 20C8 recognizes human Snail1, a 29 kDa zinc finger transcription factor. Snail1 and Slug both belong to a family of transcription factors that are responsible for inducing epithelial to mesenchymal transition (EMT) during normal organ development and tumor progression. Snail1 expression is localized to both the cytoplasm and nucleus and functions as a transcriptional repressor of E-cadherin, claudins, occludin, and desmoplakin. During embryonic development, Snail1 is localized to the mesoderm, while in the adult, expression is inducible and is usually limited to activated fibroblasts and some mesenchymal cells. Overexpression of Snail1 has been found in carcinomas and fibrosarcomas and may be used as an early marker for EMT. Applications Reported: This 20C8 antibody has been reported for use in immunohistochemical staining of formalin-fixed paraffin embedded (IHC-P) tissue, ELISA, immunoprecipitation, western blotting, and immunocytochemistry. Applications Tested: This 20C8 antibody has been tested by immunohistochemistry on formalin-fixed paraffin embedded (FFPE) human breast cancer tissue using low pH antigen retrieval. The antibody can also be used following high pH antigen retrieval. This antibody can be used at less than or equal to 5 μg/mL. It is recommended that the antibody be carefully titrated for optimal performance in the assay of interest. Purity: Greater than 90%, as determined by SDS-PAGE.
Zinc finger protein SNAI1 (SNAIL)is a zinc finger transcriptional repressor which down regulates the expression of the ectodermal gene within the mesoderm and functions to form the mesoderm and neural crest. It is reported that SNAI1 acts as an important factor of tumor invasion as evidenced by its role in E-cadherin down-regulation and induction of epithelial-mesenchymal transition (EMT). In human breast cancer, the expression of SNAI1 and/or the homologous SNAI2 (Slug) has been associated with E-cadherin repression, local or distant metastasis, tumor recurrence, or poor prognosis in different tumor series. It is also reported that Snail 1 protein in the stromamay be anew putative prognosis marker for colon tumors.Specifications
Snail1 | |
Monoclonal | |
0.5 mg/mL | |
PBS with 0.09% sodium azide; pH 7.2 | |
O95863 | |
Snai1 | |
Affinity chromatography | |
RUO | |
6615 | |
4° C | |
Liquid |
ELISA, Immunocytochemistry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
20C8 | |
Unconjugated | |
Snai1 | |
AI194338; dJ710H13.1; Protein sna; protein snail homolog 1; SLUGH2; SNA; Sna protein; Sna1; SNAH; snai; SNAI1; Snail; snail 1; snail 1 homolog; snail 1 zinc finger protein; snail 1, zinc finger protein; snail family transcriptional repressor 1; snail family zinc finger 1; snail homolog 1; Snail1; Zinc finger protein SNAI1 | |
Mouse | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG2a κ |
For Research Use Only.