Learn More
Alfa Aesar™ Smopex™-301, metal scavenger
Recovery of precious metals from catalyst processes, especially Pd, Ru, and Ir
Brand:  Alfa Aesar™ 045902.06
Code : NEW
Additional Details : Weight : 0.00500kg
Quantity 5g
 View more versions of this product
Product Code. 15412547
  • £74.30 / 5g
Estimated Shipment date
from Supplier 24-01-2018
Add to basket
Specifications
Chemical Identifiers
Smopex™-301 metal scavenger
Styryl diphenylphosphine grafted polyolefin fiber
Specifications
Smopex™-301 metal scavenger
Styryl diphenylphosphine grafted polyolefin fiber
5g
Fibers
Off-white