missing translation for 'onlineSavingsMsg'
Learn More

Smopex-101, Alfa Aesar™

For Recovery of Precious Metals From Catalyst Process Streams, Especially Pd, Rh

Brand:  Alfa Aesar 044568.14

Code : NEW

Additional Details : Weight : 0.02500kg

Quantity 25g

 View more versions of this product

Product Code. 11455340

  • £416.90 / 25g


Estimated Shipment date
from Supplier 05-02-2018
Add to basket

Specifications

Chemical Identifiers

Smopex-101
Fibers
MFCD06202258
Styrene sulfonic acid grafted polyolefin fiber

Specifications

Smopex-101
Fibers
Styrene sulfonic acid grafted polyolefin fiber
Yellow
0.3g/cm3 at 20°C
MFCD06202258
Odorless
25g
Videos
SDS