Learn More
SMER 28, Tocris Bioscience™
Positive regulator of autophagy
Brand:  RD Systems (Bio-Techne) 4297/50
Code : Z1
Additional Details : Weight : 0.00050kg
Quantity 50mg
 View more versions of this product
Product Code. 13454514
  • £474.81 / 50mg
Estimated Shipment date
from Supplier 24-03-2020
Description
Description
Positive regulator of autophagy in a mechanism independent from the mTOR pathway. Increases autophagosome synthesis and enhances clearance of model autophagy substrates such as mutant huntingtin and A53T α-synuclein, associated with Huntington's and Parkinson's diseases, respectively. Promotes reprogramming of fibroblasts to neural stem-like cells in combination with other chemical reagents.
Specifications
Chemical Identifiers
307538-42-7
264.126
smer28, smer 28, n-allyl-6-bromoquinazolin-4-amine, 6-bromo-n-2-propenyl-4-quinazolinamine, 6-bromo-n-prop-2-en-1-yl quinazolin-4-amine, 3srb, cbmicro_021891, smer26, cambridge id 6164173, smer28 2
6-bromo-N-prop-2-enylquinazolin-4-amine
C11H10BrN3
BCPOLXUSCUFDGE-UHFFFAOYSA-N
1560402
C=CCNC1=NC=NC2=C1C=C(C=C2)Br
Specifications
50mg
307538-42-7
smer28, smer 28, n-allyl-6-bromoquinazolin-4-amine, 6-bromo-n-2-propenyl-4-quinazolinamine, 6-bromo-n-prop-2-en-1-yl quinazolin-4-amine, 3srb, cbmicro_021891, smer26, cambridge id 6164173, smer28 2
C=CCNC1=NC=NC2=C1C=C(C=C2)Br
264.126
264.12
SMER 28
C11H10BrN3
BCPOLXUSCUFDGE-UHFFFAOYSA-N
6-bromo-N-prop-2-enylquinazolin-4-amine
1560402
>99%
Videos
Documentos