missing translation for 'onlineSavingsMsg'
Learn More

Smart-Vue™ Receivers

Choose from an assortment of Thermo Scientific™ Smart-Vue™ receivers including receiver packages that include power supply and software for your wireless monitoring solution.

Brand:  Thermo Scientific SV103-511-LSB

Code : NEW

Additional Details : Weight : 0.37000kg

 View more versions of this product

Product Code. 15350321

  • £1036.64 / Each

Call for latest availability
+44 (0)1509 555 500
Add to basket



USB 1.0 or 2.0 Receiver, 25mW+SW 20 sensors
Receiver, 868MHz
Velcro, CD ROM with Smart-Vue client server software 2.0 license up to 20 sensors, software drivers and user manuals.
Smart-Vue facilitates conformance to 21 CFR Part 11, cGxP and SOP compliance.
Smart-Vue™ Wireless Monitoring Solutions