Learn More
Thermo Scientific™ Smart-Vue™ Alert
Replaces the use of an analog USB model for issuing Smart-Vue wireless monitoring alerts via voice and/or SMS.
Brand:  Thermo Scientific™ SMARTVUEALERT20
Code : NEW
Additional Details : Weight : 0.00001kg
 View more versions of this product
Product Code. 15954830
  • £2813.49 / Each
Estimated Shipment: 26-01-2022
Add to basket
Description
Description
Smart-Vue™ Alert is an online alert platform, replacing the use of an analog USB modem for issuing alerts via voice and/or SMS. This is a requirement in many regions due to the phase-out of analog telephone lines. Smart-Vue Alert is a for-pay service with a yearly subscription model.
Feature summary
  • 12-month or 36-month options, time counting from the date of installation
  • Module-based license options with same ranges as server licenses: 1-20, 21-50, 51-100, 101-unlimited
  • Customer benefits from an unlimited number of alerts/calls for the allowed number of modules
  • Smart-Vue Alert supports both SMS/Text and Voice alerts
  • There is no “text-only” option (i.e. without voice alerts) at this time.
  • Users cannot customize alert messages. This feature could be studied for a future release if necessary.
Specifications
Specifications
Voice and SMS
SmartVue Server, SmartVue Client Version 2.2
Not Calibrated
Wireless Monitoring
No Display
Smart-Vue Alert, 1-20 Sensors
Smart-Vue Alert
36 month software license required to send an unlimited number of text and voice call alerts. License based on the number of sensors (dual modules count as 2 sensors). Requires SmartVueServer/SmartVue Client version 2.2 and above.