Learn More
Thermo Scientific™ Evolution™ UV-Visible Spectrophotometers Smart Thermostatted 7-Cell Rotary Changer
Efficiently control temperature with a circulating water bath using the Thermo Scientific™ Evolution™ UV-Visible Spectrophotometers Smart Thermostatted 7-Cell Rotary Changer.
Brand: Thermo Scientific™ 840-214900
Code : 04
Additional Details : Weight : 2.13300kg
Description
Efficiently control temperature with a circulating water bath using the Thermo Scientific™ Evolution™ UV-Visible Spectrophotometers Smart Thermostatted 7-Cell Rotary Changer. Combined with a liquid recirculator, this cell changer provides control of sample temperature for up to 7 samples or standards.
- Easy-to-use sample positioning accessory for use with Thermo Scientific™ Evolution™ 200 series and 300 spectrophotometers.
- Accessory control fully integrated in Thermo Scientific™ INSIGHT™ and Thermo Scientific™ VISION™ software.
- Automatic software recognition, smart alignment and serial number reporting
- Includes carousel and motor base.
- When placed in both sample and reference beams (Evolution 300 only), this cell changer allows comparison of up to 7 samples with 7 unique references.
- Monitor the temperature in a cell with Temperature probe accessory for Smart accessories, catalog number 0011401.
- When deploying this accessory in a regulated or QC environment, a full-system qualification package is available to assure compliance of instrument and accessory as a complete unit.
Specifications
Compatible with temperature probe accessory for Smart accessories, catalog#10011401 | |
Smart Thermostatted Rotary 7-Cell Changer | |
Evolution One/One Plus Series and Evolution Pro spectrophotometers | |
Thermostatting with flowing liquid from an external temperature controlled recirculator |
Evolution One/One Plus- sample position only, Evolution Pro- sample and reference positions | |
Smart Thermostatted 7-Cell Rotary Changer | |
Carousel and motor base |