Learn More
SMARCB1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA553932
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: EKENSPEKFA LKLCSELGLG GEFVTTIAYS IRGQLSWHQK TYAFSENPLP TVEIAIRNTG DADQWCPLLE TLTDAEMEKK IRDQDRNTRR MRRLANTA Highest antigen sequence identity to the following orthologs: Mouse - 100%, Rat - 100%.
SMARCB1 Polyclonal antibody specifically detects SMARCB1 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Specifications
SMARCB1 | |
Polyclonal | |
Unconjugated | |
SMARCB1 | |
AU020204; Baf47; BRG1-associated factor 47; BRG1-associated factor 47 (BAF47); CSS3; hSNF5; hSNFS; INI1; Integrase interactor 1 protein; malignant rhabdoid tumor suppressor; MRD15; mSNF5; PPP1R144; protein phosphatase 1, regulatory subunit 144; RDT; RTPS1; Sfh1p; SMARCB1; smarcb1a; SNF5; SNF5 homolog; SNF5/INI1; SNF5L1; Snr1; Sucrose nonfermenting yeast homolog like 1; sucrose nonfermenting, yeast, homolog-like 1; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily b, member 1; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily b, member 1 isoform a; SWI/SNF related, matrix associated, actin dependent regulator of chromatin, subfamily b, member 1a; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily B member 1; SWI/SNF-related matrix-associated actin-dependent regulator of chromatin subfamily B member 1-A; SWI/SNF-related matrix-associated protein; SWI10; SWNTS1; Transcription factor TYE | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
20587, 361825, 6598 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q12824, Q9Z0H3 | |
SMARCB1 | |
Recombinant protein corresponding to Human SMARCB1 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Research Use Only