Learn More
Hamilton™ Stainless Steel Small Hub Removable Custom Needles
Used for animal injections to limit tissue damage and sample loss.
£146.00 - £320.00
Products 7
Product Code Brand Needle Gauge Price Quantity & Availability  
Product Code Brand Needle Gauge Price Quantity & Availability  
12311583
View Documents Promotion Details
Hamilton™
7804-09
23s G
£146.00
Pack of 6
View alternative products
Save Up to 
»
Estimated Shipment: 21-06-2024
to see stock.
10509822
View Documents Promotion Details
Hamilton™
207419
19 G
£160.00
Pack of 6
View alternative products
Save Up to 
»
Estimated Shipment: 21-06-2024
to see stock.
12396289
View Documents Promotion Details
Hamilton™
7731-06
25s G
£320.00
Pack of 6
View alternative products
Save Up to 
»
Estimated Shipment: 21-06-2024
to see stock.
12376439
View Documents Promotion Details
Hamilton™
7804-05
25 G
£150.00
Pack of 6
View alternative products
Save Up to 
»
Estimated Shipment: 13-05-2024
to see stock.
12326449
View Documents Promotion Details
Hamilton™
7804-11
20 G
£146.00
Pack of 6
View alternative products
Save Up to 
»
Estimated Shipment: 15-05-2024
to see stock.
12336299
View Documents Promotion Details
Hamilton™
7731-10
20 G
£320.00
Pack of 6
View alternative products
Save Up to 
»
Estimated Shipment: 21-06-2024
to see stock.
11512525
View Documents Promotion Details
Hamilton™
207419
19 G This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A View alternative products
Save Up to 
»
Description
Description
  • Autoclavable
  • Provided in packs of 6
  • Can be used with removable (RN)-style syringes that are 100μL and smaller
  • Custom lengths and point styles are available
  • Removable needles have minimal dead volume and for this reason are recommended over a luer or luer lock connection