missing translation for 'onlineSavingsMsg'
Learn More

SLITRK4 Goat anti-Human, Polyclonal, R&D Systems™

Goat Polyclonal Antibody

Brand:  RD Systems (Bio-Techne) AF2860

Code : Z1

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 13447834

  • £352.24 / 50µg

Estimated Shipment
13-08-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

SLITRK4 Polyclonal antibody AF2860 Specifically detects SLITRK4 in Human samples. It is validated for Western Blotting
Specifications

Specifications

SLITRK4
LYOPH
Western Blot 0.1 ug/mL
Q8IW52
DKFZp547M2010, SLIT and NTRK-like family, member 4, SLIT and NTRK-like protein 4, slit and trk like gene 4
Goat
IgG
50 ug
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Polyclonal
Detects human SLITRK4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, less than 5% cross-reactivity with recombinant human (rh) SLITRK1, rhSLITRK2, and rhSLITRK5 is observed.
Human
Western Blot
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Affinity Purified
SLITRK4
Mouse myeloma cell line NS0-derived recombinant human SLITRK4 Asp19-Pro616 Accession # Q8IW52
Immunogen affinity purified
RUO
Primary
139065
Reconstitute at 0.2 mg/mL in sterile PBS.
Videos
Documents

For Research Use Only