missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLITRK2 Goat anti-Human, Polyclonal, R&D Systems™
Goat Polyclonal Antibody
POA
Specifications
Antigen | SLITRK2 |
---|---|
Immunogen | Mouse myeloma cell line NS0-derived recombinant human SLITRK2 Thr19-Pro617 Accession # Q9H156 |
Gene ID (Entrez) | 84631 |
Gene Accession No. | Q9H156 |
Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
Description
SLITRK2 Polyclonal specifically detects SLITRK2 in Human samples. It is validated for Western Blot.Specifications
SLITRK2 | |
84631 | |
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
Primary | |
Detects human SLITRK2 in direct ELISAs and Western blots. In direct ELISAs, approximately 10% cross-reactivity with recombinant human (rh) SLITRK4 is observed and less than 2% cross-reactivity with rhSLITRK1 and rhSLITRK5 is observed. | |
Unconjugated | |
Western Blot | |
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Mouse myeloma cell line NS0-derived recombinant human SLITRK2 Thr19-Pro617 Accession # Q9H156 | |
Q9H156 | |
Reconstitute at 0.2 mg/mL in sterile PBS. | |
Goat | |
IgG | |
Affinity Purified | |
RUO |
For Research Use Only