Learn More
SLCO2B1 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA554258
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: GITHQTSAHP GLELSPSCME ACSCPLDGFN PVCDPSTRVE YITPCHAGCS SWVVQDALDN SQVFYTNCSC VVEGNPVLAG SCDSTCSHLV VPF Highest antigen sequence identity to the following orthologs: Mouse - 63%, Rat - 63%.
SLCO2B1 Polyclonal antibody specifically detects SLCO2B1 in Human samples. It is validated for Immunohistochemistry.Specifications
SLCO2B1 | |
Polyclonal | |
Unconjugated | |
SLCO2B1 | |
AI060904; AI852653; hypothetical protein LOC557480; im:7158298; KIAA0880; moat1; multispecific organic anion transporter 1; Oatp2b1; OATPB; OATP-B; OATP-RP2; Organic anion transporter B; organic anion transporter moatp1; Organic anion transporter polypeptide-related protein 2; organic anion transporting polypeptide 2b1; organic anion-transporting polypeptide 2B1; SLC21A9; SLCO/OATP2B1; Slco2b1; solute carrier anion transporting family member 2b1; solute carrier family 21 (organic anion transporter), member 9; solute carrier family 21 member 9; Solute carrier organic anion transporter family member 2B1; solute carrier organic anion transporter family, member 2B1; wu:fi40d02; zgc:123236 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
11309 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O94956 | |
SLCO2B1 | |
Recombinant protein corresponding to Human SLCO2B1 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only