missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC38A11 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA560362
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: AISLGPHIPK TEDAWVFAKP NAIQ Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 79%.
SLC38A11 Polyclonal antibody specifically detects SLC38A11 in Human samples. It is validated for Immunohistochemistry.Specifications
SLC38A11 | |
Polyclonal | |
Unconjugated | |
SLC38A11 | |
AVT2; Putative sodium-coupled neutral amino acid transporter 11; SLC38A11; Solute carrier family 38 member 11; solute carrier family 38, member 11 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
151258 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q08AI6 | |
SLC38A11 | |
Recombinant protein corresponding to Human SLC38A11 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Research Use Only