missing translation for 'onlineSavingsMsg'
Learn More
Learn More
SLC25A12 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Thermo Scientific PA557233
Additional Details : Weight : 0.02000kg
Description
Immunogen sequence: FNWDCEFIRL HFGHNRKKHL NYTEFTQFLQ ELQLEHARQA FALKDKSKSG MISGLDFSDI MV Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
SLC25A12 Polyclonal antibody specifically detects SLC25A12 in Human, Mouse, Rat samples. It is validated for Immunohistochemistry, Western Blot.Specifications
SLC25A12 | |
Polyclonal | |
Unconjugated | |
SLC25A12 | |
2610002D09Rik; AGC1; AI839531; araceli hiperlarga; ARALAR; Aralar1; B230107K20Rik; BB129864; calcium binding mitochondrial carrier superfamily member Aralar1; calcium-binding mitochondrial carrier protein Aralar1; Calcium-binding mitochondrial carrier protein Aralar1-like protein; mitochondrial aspartate glutamate carrier 1; RGD1561141; SLC25A12; solute carrier family 25 (aspartate/glutamate carrier), member 12; solute carrier family 25 (mitochondrial carrier, Aralar), member 12; solute carrier family 25 member 12; solute carrier family 25, member 12 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
362145, 78830, 8604 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunohistochemistry, Western Blot | |
0.1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
O75746, Q8BH59 | |
SLC25A12 | |
Recombinant protein corresponding to Human SLC25A12 | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Research Use Only