Learn More
SAP (SLAM-Associated Protein) Monoclonal Antibody (XLP-1D12), eFluor™ 660, eBioscience™, Invitrogen™
Rat Monoclonal Antibody
£116.00 - £297.00
Specifications
Antigen | SAP (SLAM-Associated Protein) |
---|---|
Clone | XLP-1D12 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | eFluor 660 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
15570137
|
Affymetrix eBioscience
50-9787-42 |
100 Tests |
£297.00
100 tests |
Estimated Shipment: 24-05-2024 Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | ||||
15560137
|
Affymetrix eBioscience
50-9787-41 |
25 Tests |
This item is currently unavailable or has been discontinued. View the product page for possible alternatives. |
||||||
Description
This gene encodes a protein that plays a major role in the bidirectional stimulation of T and B cells. This protein contains an SH2 domain and a short tail. It associates with the signaling lymphocyte-activation molecule, thereby acting as an inhibitor of this transmembrane protein by blocking the recruitment of the SH2-domain-containing signal-transduction molecule SHP-2 to its docking site. This protein can also bind to other related surface molecules that are expressed on activated T, B and NK cells, thereby modifying signal transduction pathways in these cells. Mutations in this gene cause lymphoproliferative syndrome X-linked type 1 or Duncan disease, a rare immunodeficiency characterized by extreme susceptibility to infection with Epstein-Barr virus, with symptoms including severe mononucleosis and malignant lymphoma. Multiple transcript variants encoding different isoforms have been found for this gene.
SAP (SLAM-Associated Protein) Monoclonal antibody specifically detects SAP (SLAM-Associated Protein) in Human samples. It is validated for Flow Cytometry.Specifications
SAP (SLAM-Associated Protein) | |
Flow Cytometry | |
eFluor 660 | |
Rat | |
Human | |
4068 | |
Primary | |
Antibody |
XLP-1D12 | |
Monoclonal | |
Liquid | |
RUO | |
O60880 | |
IgG2a κ | |
Affinity chromatography |
For Research Use Only.