Learn More
SKF 97541, Tocris Bioscience™
Extremely potent GABAB agonist
Brand:  Tocris (Bio-Techne) 037950
Code : Z1
Additional Details : Weight : 0.01000kg
Quantity 50mg
 View more versions of this product
Product Code. 13487593
  • £626.00 / 50mg
Estimated Shipment
02-03-2018
Add to basket
Description
Description
Very potent GABAB agonist, at least ten times more active than baclofen (Cat. No. 0417). Also GABAA-ρ antagonist.
Specifications
Chemical Identifiers
SKF 97541
C4H12NO2P
137.12
3-Aminopropyl(methyl)phosphinic acid
Specifications
SKF 97541
>98%
C4H12NO2P
3-Aminopropyl(methyl)phosphinic acid
127729-35-5
50mg
137.12
Videos