missing translation for 'onlineSavingsMsg'
Learn More

SKF 96365 hydrochloride, Tocris Bioscience™

TRPC channel blocker; also inhibits store-operated Ca2+ entry

Brand:  Tocris (Bio-Techne) 114750

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13468733

  • £717.36 / 50mg

Estimated Shipment date
from Supplier 30-03-2020

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Store-operated Ca2+ entry (SOCE) inhibitor that inhibits STIM1. Also blocks TRPC channels, voltage-gated Ca2+ channels and potassium channels.
Specifications

Chemical Identifiers

130495-35-1
402.919
skf 96365 hydrochloride, skf-96365 hydrochloride, skf-96365, hydrochloride, 1-2-3-4-methoxyphenyl propoxy-4-methoxyphenylethyl-1h-imidazole hydrochloride, 1-2-3-4-methoxyphenyl propoxy-4-methoxyphenylethyl-1h-imidazole, 1-beta-3-4-methoxyphenyl propoxy-4-methoxyphenethyl-1h-imidazole, hcl, 1-2-4-methoxyphenyl-2-3-4-methoxyphenyl propoxy ethyl-1h-imidazole hydrochloride, 1-2-4-methoxyphenyl-2-3-4-methoxyphenyl propoxy ethyl imidazole hydrochloride, 1h-imidazole, 1-2-4-methoxyphenyl-2-3-4-methoxyphenyl propoxy ethyl-, monohydrochloride
1-[2-(4-methoxyphenyl)-2-[3-(4-methoxyphenyl)propoxy]ethyl]imidazole;hydrochloride
C22H27ClN2O3
FWLPKVQUECFKSW-UHFFFAOYSA-N
104955
COC1=CC=C(C=C1)CCCOC(CN2C=CN=C2)C3=CC=C(C=C3)OC.Cl

Specifications

50mg
130495-35-1
skf 96365 hydrochloride, skf-96365 hydrochloride, skf-96365, hydrochloride, 1-2-3-4-methoxyphenyl propoxy-4-methoxyphenylethyl-1h-imidazole hydrochloride, 1-2-3-4-methoxyphenyl propoxy-4-methoxyphenylethyl-1h-imidazole, 1-beta-3-4-methoxyphenyl propoxy-4-methoxyphenethyl-1h-imidazole, hcl, 1-2-4-methoxyphenyl-2-3-4-methoxyphenyl propoxy ethyl-1h-imidazole hydrochloride, 1-2-4-methoxyphenyl-2-3-4-methoxyphenyl propoxy ethyl imidazole hydrochloride, 1h-imidazole, 1-2-4-methoxyphenyl-2-3-4-methoxyphenyl propoxy ethyl-, monohydrochloride
COC1=CC=C(C=C1)CCCOC(CN2C=CN=C2)C3=CC=C(C=C3)OC.Cl
402.919
402.92
SKF 96365 hydrochloride
C22H27ClN2O3
FWLPKVQUECFKSW-UHFFFAOYSA-N
1-[2-(4-methoxyphenyl)-2-[3-(4-methoxyphenyl)propoxy]ethyl]imidazole;hydrochloride
104955
>99%
Videos
Documents