missing translation for 'onlineSavingsMsg'
Learn More

SKF 86466 hydrochloride, Tocris Bioscience™

Potent and selective α2 antagonist

Brand:  Tocris (Bio-Techne) 386610

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13463724

  • £124.20 / 10mg


Estimated Shipment
28-03-2019
Add to basket

Description

Description

Potent and selective α 2 antagonist at pre- and post-junctional α 2 -adrenoceptors (K i values are 13 and 17 nM respectively). Exhibits antihypertensive activity in a rat model.
Specifications

Chemical Identifiers

86129-54-6
232.148
skf-86466 hydrochloride, skf 86466 hydrochloride, 6-chloro-2,3,4,5-tetrahydro-3-methyl-1h-3-benzazepine hydrochloride, 6-chloro-n-methyl-2,3,4,5-tetrahydro-1-h-3-benzazepine hydrochloride, 1h-3-benzazepine, 6-chloro-2,3,4,5-tetrahydro-3-methyl-, hydrochloride, 6-chloro-3-methyl-2,3,4,5-tetrahydro-1h-benzo d azepine hydrochloride, benalfocin hydrochloride, skf-86466 hydrochloride, solid hplc
6-chloro-3-methyl-1,2,4,5-tetrahydro-3-benzazepine;hydrochloride
C11H15Cl2N
JKQKVNMNAIFCFS-UHFFFAOYSA-N
10220250
CN1CCC2=C(CC1)C(=CC=C2)Cl.Cl

Specifications

SKF 86466 hydrochloride
86129-54-6
skf-86466 hydrochloride, skf 86466 hydrochloride, 6-chloro-2,3,4,5-tetrahydro-3-methyl-1h-3-benzazepine hydrochloride, 6-chloro-n-methyl-2,3,4,5-tetrahydro-1-h-3-benzazepine hydrochloride, 1h-3-benzazepine, 6-chloro-2,3,4,5-tetrahydro-3-methyl-, hydrochloride, 6-chloro-3-methyl-2,3,4,5-tetrahydro-1h-benzo d azepine hydrochloride, benalfocin hydrochloride, skf-86466 hydrochloride, solid hplc
CN1CCC2=C(CC1)C(=CC=C2)Cl.Cl
232.148
232.15
10mg
C11H15Cl2N
JKQKVNMNAIFCFS-UHFFFAOYSA-N
6-chloro-3-methyl-1,2,4,5-tetrahydro-3-benzazepine;hydrochloride
10220250
>98%
Videos