missing translation for 'onlineSavingsMsg'
Learn More

SKF 83566 hydrobromide, Tocris Bioscience™

Potent, selective D1-like antagonist

Brand:  Tocris (Bio-Techne) 158650

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 50mg

 View more versions of this product

Product Code. 13439723

  • £368.29 / 50mg

Estimated Shipment
27-06-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Potent and selective D1-like dopamine receptor antagonist (Ki∽ 0.56 nM for D1; KB = 2 μM for D2). Also antagonist at the vascular 5-HT2 receptor (Ki = 11 nM). Displays selective inhibition of adenylyl cyclase 2 (AC2); inactive against AC1 or AC5. Centrally active following systemic administration in vivo.
Specifications

Chemical Identifiers

108179-91-5
413.153
skf 83566 hydrobromide, 8-bromo-3-methyl-5-phenyl-2,3,4,5-tetrahydro-1h-benzo d azepin-7-ol hydrobromide, 8-bromo-2,3,4,5-tetrahydro-3-methyl-5-phenyl-1h-3-benzazepin-7-ol hydrobromide, 8-bromo-2,3,4,5-tetrahydro-3-methyl-5-phenyl-1h-3-benzazepin-7-olhydrobromide, 8-bromo-3-methyl-5-phenyl-1,2,4,5-tetrahydro-3-benzazepin-7-ol hydrobromide, 8-bromo-3-methyl-5-phenyl-2,3,4,5-tetrahydro-1h-3-benzazepin-7-ol hydrobromide
8-bromo-3-methyl-5-phenyl-1,2,4,5-tetrahydro-3-benzazepin-7-ol;hydrobromide
C17H19Br2NO
SDQJYYGODYRPBR-UHFFFAOYSA-N
23581817
CN1CCC2=CC(=C(C=C2C(C1)C3=CC=CC=C3)O)Br.Br

Specifications

SKF 83566 hydrobromide
108179-91-5
skf 83566 hydrobromide, 8-bromo-3-methyl-5-phenyl-2,3,4,5-tetrahydro-1h-benzo d azepin-7-ol hydrobromide, 8-bromo-2,3,4,5-tetrahydro-3-methyl-5-phenyl-1h-3-benzazepin-7-ol hydrobromide, 8-bromo-2,3,4,5-tetrahydro-3-methyl-5-phenyl-1h-3-benzazepin-7-olhydrobromide, 8-bromo-3-methyl-5-phenyl-1,2,4,5-tetrahydro-3-benzazepin-7-ol hydrobromide, 8-bromo-3-methyl-5-phenyl-2,3,4,5-tetrahydro-1h-3-benzazepin-7-ol hydrobromide
CN1CCC2=CC(=C(C=C2C(C1)C3=CC=CC=C3)O)Br.Br
413.153
413.15
50mg
C17H19Br2NO
SDQJYYGODYRPBR-UHFFFAOYSA-N
8-bromo-3-methyl-5-phenyl-1,2,4,5-tetrahydro-3-benzazepin-7-ol;hydrobromide
23581817
>98%
Videos