Learn More
Tocris Bioscience™ SKA 31
Activator of KCa3.1 and KCa2 channels
Brand:  Tocris Bioscience™ 367050
Code : Z1
Additional Details : Weight : 0.01000kg
 View more versions of this product
Product Code. 13443394
  • £408.22 / 50mg
Estimated Shipment date
from Supplier 24-09-2019
Description
Description
Activator of KCa3.1 and KCa2 channels (EC50 values are 260, 2900, 2900 nM for KCa3.1, KCa2.1 and KCa2.2 respectively). Potentiates acetylcholine-induced EDHF-type dilations of mouse carotid arteries and lowers blood pressure in normotensive and hypertensive mice.
Specifications
Specifications
SKA 31
>99
50mg
SKA 31
200.26