Learn More
Tocris Bioscience™ SKA 31
Activator of KCa3.1 and KCa2 channels
Brand:  Tocris Bioscience™ 367010
Code : Z1
Additional Details : Weight : 0.01100kg
 View more versions of this product
Product Code. 13433394
  • £139.65 / 10mg
Estimated Shipment date
from Supplier 24-03-2020
Description
Description
Activator of KCa3.1 and KCa2 channels (EC50 values are 260, 2900, 2900 nM for KCa3.1, KCa2.1 and KCa2.2 respectively). Potentiates acetylcholine-induced EDHF-type dilations of mouse carotid arteries and lowers blood pressure in normotensive and hypertensive mice.
Specifications
Specifications
SKA 31
>99
10mg
SKA 31
200.26