missing translation for 'onlineSavingsMsg'
Learn More

(+)-SK&F 10047 hydrochloride, Tocris Bioscience™

σ1 selective agonist

Brand:  RD Systems (Bio-Techne) 1079/50

Code : Z1

Additional Details : Weight : 0.00050kg

Quantity 50mg

 View more versions of this product

Product Code. 13448513

  • £786.45 / 50mg

Call for latest availability
+44 (0)1509 555 500

Please to purchase this item. Need a web account? Register with us today!


Description

Description

Prototypical σ1 receptor agonist. Behaviorally active in animal models of memory and stress.
Specifications

Chemical Identifiers

133005-41-1
293.835
+-SK&F 10047 hydrochloride, 2s,6s,11s-1,2,3,4,5,6-hexahydro-6,11-dimethyl-3-2-propenyl-2,6-methano-3-benzazocin-8-ol hydrochloride
CC1C2CC3=C(C1(CCN2CC=C)C)C=C(C=C3)O.Cl
C17H24ClNO
ZTGMHFIGNYXMJV-XXZNMZKQSA-N
121513842

Specifications

50mg
133005-41-1
+-SK&F 10047 hydrochloride, 2s,6s,11s-1,2,3,4,5,6-hexahydro-6,11-dimethyl-3-2-propenyl-2,6-methano-3-benzazocin-8-ol hydrochloride
CC1C2CC3=C(C1(CCN2CC=C)C)C=C(C=C3)O.Cl
121513842
>98%
(+)-SK&F 10047 hydrochloride
C17H24ClNO
ZTGMHFIGNYXMJV-XXZNMZKQSA-N
293.835
293.84
Videos
Documents