missing translation for 'onlineSavingsMsg'
Learn More

SJ 172550, Tocris Bioscience™

MDMX inhibitor. Disrupts MDMX-p53 interaction

Brand:  Tocris (Bio-Techne) 392910

Code : Z1

Additional Details : Weight : 0.01000kg

Quantity 10mg

 View more versions of this product

Product Code. 13433824

  • £180.81 / 10mg

Estimated Shipment date
from Supplier 17-12-2019

Please to purchase this item. Need a web account? Register with us today!


Description

Description

MDMX inhibitor. Reversibly binds MDMX (EC50 = 2.3 μM). Inhibits the MDMX-p53 interaction in cultured retinoblastoma cells; frees p53 to induce apoptosis.
Specifications

Chemical Identifiers

431979-47-4
428.869
methyl 2-2-chloro-6-ethoxy-4-3-methyl-5-oxo-1-phenylpyrazol-4-ylidene methyl phenoxy acetate
methyl 2-[2-chloro-6-ethoxy-4-[(3-methyl-5-oxo-1-phenylpyrazol-4-ylidene)methyl]phenoxy]acetate
C22H21ClN2O5
RKKFQJXGAQWHBZ-UHFFFAOYSA-N
2913564
CCOC1=C(C(=CC(=C1)C=C2C(=NN(C2=O)C3=CC=CC=C3)C)Cl)OCC(=O)OC

Specifications

10mg
431979-47-4
methyl 2-2-chloro-6-ethoxy-4-3-methyl-5-oxo-1-phenylpyrazol-4-ylidene methyl phenoxy acetate
CCOC1=C(C(=CC(=C1)C=C2C(=NN(C2=O)C3=CC=CC=C3)C)Cl)OCC(=O)OC
428.869
428.87
SJ 172550
C22H21ClN2O5
RKKFQJXGAQWHBZ-UHFFFAOYSA-N
methyl 2-[2-chloro-6-ethoxy-4-[(3-methyl-5-oxo-1-phenylpyrazol-4-ylidene)methyl]phenoxy]acetate
2913564
>99%
Videos
Documents