missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ VNN1 Recombinant Mouse Protein, hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the mouse VNN1 (Q9Z0K8) (Met 1-Ser 487) without the pro peptides

Brand:  Thermo Scientific™ 50952M02H25

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15389316

  • £245.68 / 25µg


Estimated Shipment
15-06-2018
Add to basket

Includes: Five vials of protein in suspension buffer

Description

Description

Vanin 1 (VNN1), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • This protein has a Fc region of human IgG1 at the C-terminus
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Activity: The activity of this protein has not been tested
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
22361
PAGE
5 x 5μg
C-Terminal
<1 EU/μg
Vanin 1
Assay Standard
Mammalian
Lyophilized
79kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Full Length/Recombinant
V-1
VNN1
Other Proteins
IgG1 Fc Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.