Sino Biological™ VEGFC Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the mature form of human VEGFC (NP_005420.1), residues 103 to 227, fused to a polyhistidine tag at the C-terminus

Overview
Brand: Invitrogen

Manufacturer Part Number: 10542H08H25

VEGFC Recombinant Human Protein (His Tag, Active),25 µg

UNSPSC: 12352202

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 13485896

Quantity Price
1 £ 184.0 / 25µg
Estimated Shipment date
from Supplier 07-02-2017
Add to basket


Description and Specification

Specification

For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Molecular Weight 15.5kDa
Purity ≥90% by SDS-PAGE
Quantity 5 x 5μg
Storage Requirements Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Structural Form Recombinant
Protein Subtype VEGF (Vascular Endothelial Growth Factor)
Endotoxin Concentration <1 EU/μg
Gene Alias Flt4-L, VRP, VEGFC
Common Name Vascular endothelial growth factor C
Gene Symbol VEGFC
Protein Tag His Tag
Validated Application Assay Standard
Protein Family Growth Factors & Receptors
Expression System Mammalian
Format Lyophilized
Gene ID (Entrez) 7424

Vascular endothelial growth factor C (VEGFC), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; Scatchard analysis showed the affinity constant (Kd) of recombinant human VEGFC bound to recombinant human VEGFR3 was 1.4nM
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.