missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ VCAM1 Recombinant Human Protein, N-term His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain of human VCAM1 (NP_001069.1) (Phe 25 - Pro 697) fused to a polyhistidine tag at the N-terminus

Brand:  Thermo Scientific™ 10113H07H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15224665

  • £183.22 / 50µg


Estimated Shipment
10-07-2018
Add to basket

Description

Description

Vascular cell adhesion molecule 1 (VCAM1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure, and performing cell based assays. It can also be used as an immunogen, as a protein standard, or in other research applications.
  • Activity: Measured by the ability of the immobilized protein to support the adhesion of U937 human histiocytic lymphoma cells; when cells are added to VCAM1 coated plates (10μg/mL, 100μL/well ) approximately 85-95% will adhere specifically
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
76kDa
50μg
Truncated/Recombinant/Cell Surface Protein
<1 EU/μg
Vascular cell adhesion molecule 1
Assay Standard
Mammalian
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Other Proteins
VCAM1, CD106, DKFZp779G2333, INCAM-100, mgC99561
VCAM1
CD Proteins
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.