missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ USP5 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human USP5 isoform short (P45974-2) (Met 1-Ser 835) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 12772H08B25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15236055

  • £127.01 / 25µg


Estimated Shipment
07-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Ubiquitin Carboxyl-Terminal Hydrolase 5 (USP5), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 50mM Tris, 100mM NaCl, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
8078
PAGE
5 x 5μg
C-Terminal
Proteases
ISOT
USP5
Proteases, Peptidases & Inhibitors
His Tag
Lyophilized
94.7kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Full Length/Recombinant
<1 EU/μg
Ubiquitin carboxyl-terminal hydrolase 5
Assay Standard
Insect
Videos