missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ UNG Recombinant Human Protein, N-GST Tag

This recombinant protein was expressed from a DNA sequence encoding the human UNG isoform 1 (P13051-2) (Phe 85-Leu 304) fused to the GST tag at the N-terminus

Brand:  Thermo Scientific™ 12939H09E25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15296085

  • £233.19 / 25µg


Estimated Shipment
09-03-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Description

Description

Uracil-DNA Glycosylase (UNG), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in 40mM Tris, 150mM NaCl, 2mM GSH, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
7374
PAGE
5 x 5μg
N-Terminal
<1 EU/μg
Uracil-DNA glycosylase
Assay Standard
E. coli
Lyophilized
52kDa
≥85% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Full Length/Recombinant
DGU, DKFZp781L1143, HIGM4, UDG, UNG1, UNG15, UNG2
UNG
Mitochondrial Proteins
GST Tag
Videos