missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ UCHL3 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human UCHL3 (NP_005993.1) (Glu 2-Ala 230) fused to a polyhistide tag at the N-terminus

Brand:  Thermo Scientific™ 11634H07E5

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15238755

  • £93.43 / 5µg


Estimated Shipment
11-05-2018
Add to basket

Description

Description

Ubiquitin Carboxyl-Terminal Hydrolase Isozyme L3 (UCHL3), recombinant human protein is supplied in frozen format. It is suitable for use in protein studies such as enzymatic analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its ability to hydrolyze Ubiquitin AMC; the specific activity is >14,000pmole/min./μg
  • Formulation: 20mM Tris, 500mM NaCl, 0.2μg/μL BSA, 20%glycerol, 1mM DTT pH 8.0
  • Reconstitution: No reconstitution needed

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
27kDa
5μg
N-Terminal
RP11-173B14.3, UCH-L3, UCHL3
UCHL3
E. coli
Frozen
≥90% by SDS-PAGE
Store at -80°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
Proteases
Ubiquitin carboxyl-terminal hydrolase isozyme L3
Assay Standard
His Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.