Sino Biological™ TREML2 Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the human TREML2 (NP_079083.2) extracellular domain (Met 1-Ser 268) fused to a polyhistidine tag at the C-terminus

Overview
Brand: Invitrogen

Manufacturer Part Number: 11655H08H5

TREML2 RECOMBINANT HUMAN PROTEIN, HIS TAG

UNSPSC: 12352200

Additional Details:
Additional Details: Weight: 0.01000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 15208795

Quantity Price
1 £ 55.0 / 5µg
Estimated Shipment date
from Supplier 09-03-2017
Add to basket


Description and Specification

Specification

Protein Subtype Other Proteins
Gene Alias UNQ6268/PRO20473, C6orf76, FLJ13693, mgC149715, mgC149716,TLT-2, TLT2, dJ238O23.1
Protein Tag His Tag
Expression System Mammalian
Molecular Weight 28.5kDa
Purification Method PAGE
Purity ≥90% by SDS-PAGE
Quantity 5μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Termination C-Terminal
Structural Form Truncated/Recombinant
Endotoxin Concentration <1 EU/μg
Common Name Triggering receptor expressed on myeloid cells-like 2
Gene Symbol TREML2
Validated Application Assay Standard
Protein Family Other Proteins
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene ID (Entrez) 79865

Triggering Receptor Expressed on Myeloid Cells Like 2 (TREML2), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.