Learn More
TREM2 Recombinant Mouse Protein, His Tag, Invitrogen™ Sino Biological™
This recombinant protein was expressed from a DNA sequence encoding the mouse TREM2 (NP_112544.1) extracellular domain (Met 1-Pro 168) fused to a C-terminal polyhistidine tag
Brand: Invitrogen 50149M08H5
Code : NEW
Additional Details : Weight : 0.01000kg
Includes: One vial of protein in suspension buffer
Description
Triggering Receptor Expressed on Myeloid Cells 2 (TREM2), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.- Activity: The activity of this protein has not been tested
- Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
- Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
This protein is manufactured by Sino Biological Inc.
Specifications
Protein Expression and Isolation, Enzyme and Protein Activity Assay, Protein Assay Control, Reference Standards and Accessories | |
18kDa | |
5μg | |
C-Terminal | |
<1 EU/μg | |
Triggering receptor expressed on myeloid cells 2 | |
His Tag | |
Other Proteins | |
≥95% by SDS-PAGE |
83433 | |
PAGE | |
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. | |
Truncated/Recombinant | |
Trem2a, Trem2b, Trem2c | |
TREM2 | |
Mammalian | |
Lyophilized |
For Research Use Only. Not for use in diagnostic procedures.