missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ TREM1 Recombinant Human Protein, hIgG1-Fc.His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Arg 200) of human TREM1 (NP_061113.1) precursor fused to the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 10511H03H250

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15276845

  • £732.87 / 250µg


Estimated Shipment
10-05-2018
Add to basket

Description

Description

Triggering Receptor Expressed on Myeloid Cells 1 (TREM1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
54210
PAGE
5 x 50μg
C-Terminal
<1 EU/μg
Triggering receptor expressed on myeloid cells 1
Assay Standard
Mammalian
Lyophilized
48.3kDa
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Truncated/Recombinant
TREM-1, TREM1
TREM1
Other Proteins
IgG1 Fc Tag
Videos

For Research Use Only. Not for use in diagnostic procedures.