Sino Biological™ TREM1 Recombinant Human Protein, hIgG1-Fc.His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Arg 200) of human TREM1 (NP_061113.1) precursor fused to the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus

Overview
Brand: Invitrogen

Manufacturer Part Number: 10511H03H25

TREM1 RECOMBINANT HUMAN PROTEIN, HIGG1-FC.HIS TAG

UNSPSC: 12352200

Code: NEW

Additional Details:
Additional Details: Weight: 0.01000kg



Disclaimers: For Research Use Only. Not for use in diagnostic procedures.

Product Code. 15266845

Quantity Price
1 £ 119.0 / 25µg
EU Stock 1
Add to basket


Description and Specification

Specification

Molecular Weight 48.3kDa
Purification Method PAGE
Purity ≥90% by SDS-PAGE
Quantity 5 x 5μg
Storage Requirements Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles.
Termination C-Terminal
Structural Form Truncated/Recombinant
Protein Subtype Other Proteins
Endotoxin Concentration <1 EU/μg
Gene Alias TREM-1, TREM1
Common Name Triggering receptor expressed on myeloid cells 1
Gene Symbol TREM1
Protein Tag IgG1 Fc Tag
Validated Application Assay Standard
Protein Family Other Proteins
Expression System Mammalian
For Use With (Application) Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
Format Lyophilized
Gene ID (Entrez) 54210

Triggering Receptor Expressed on Myeloid Cells 1 (TREM1), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: The activity of this protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.