missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ TNFSRF9 Recombinant Human Protein, hIgG1-Fc.His Tag

This recombinant protein was expressed from a DNA sequence encoding the N-terminal fragment (Met 1-Gln 186) of the extracellular domain of human TNFSRF9 (NP_001552.2) fused with the C-terminal polyhistidine-tagged Fc region of human IgG1 at the C-terminus

Brand:  Thermo Scientific™ 10041H03H250

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15244525

  • £471.58 / 250µg


Estimated Shipment
15-06-2018
Add to basket

Description

Description

Tumor Necrosis Factor Receptor Super Family 9 (TNFSRF9), recombinant human/mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant mouse 4-1BB Ligand at 20μg/mL (100μL/well) can bind human 4-1BB with a linear range of 15.6-500ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
45.2kDa
5 x 50μg
TNF Receptors
4-1BB, CD137, CDw137, ILA, mgC2172, TNFSRF9
TNFSRF9
Cytokines & Receptors/CD Proteins
His Tag/IgG1 Fc Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Tumor necrosis factor receptor superfamily, member 9
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.