missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ TNFSRF9 (CD137) Recombinant Human Protein, His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Met 1-Gln 186) of human TNFSRF9 (NP_001552.2) fused to a polyhistidine tag at the C-terminus

Brand:  Thermo Scientific™ 10041H08H50

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 13455666

  • £236.31 / 50µg


Estimated Shipment
15-06-2018
Add to basket

Description

Description

Tumor necrosis factor receptor superfamily, member 9 (TNFSRF9/CD137), recombinant human protein is supplied as a lyophilized powder. It is suitable for use in analysis of protein structure. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
Activity: This recombinant protein has not been tested
  • Formulation: Lyophilized in PBS, pH 7.4, 5% Mannitol, 5% Trehalose, and 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible
  • Shipping Condition: Room Temperature. This protein is manufactured by Sino Biological Inc.

    Specifications

    Specifications

    Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
    3604
    ≥85% by SDS-PAGE
    Store at -20° to -70°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
    TNF (Tumor Necrosis Factor)
    4-1BB, CD137, CDw137, ILA, mgC2172, TNFSRF9
    TNFSRF9
    Cytokines & Receptors/CD Proteins
    His Tag
    Lyophilized
    18.8kDa
    50μg
    Truncated/Recombinant
    <1 EU/μg
    Tumor necrosis factor receptor superfamily, member 9
    Assay Standard
    Mammalian
    Videos

    For Research Use Only. Not for use in diagnostic procedures.