missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ TNFSF9 Recombinant Mouse Protein, N-His Tag

This recombinant protein was expressed from a DNA sequence encoding the extracellular domain (Arg 104-Glu 309) of the mouse TNFSF9 precursor (NP_033430.1) fused to a polyhistidine tag at the N-terminus

Brand:  Thermo Scientific™ 50067M07H25

Code : NEW

Additional Details : Weight : 0.01000kg

 View more versions of this product

Product Code. 15332636

  • £226.26 / 25µg


Estimated Shipment
05-02-2018
Add to basket

Disclaimers:

For Research Use Only. Not for use in diagnostic procedures.


Includes: Five vials of lyophilized protein

Description

Description

Tumor necrosis factor (ligand) superfamily, member 9 (TNFSF9), recombinant mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant mouse TNFSF9 at 20μg/mL (100μL/well) can bind human TNFSRF9 with a linear range of 15.6-500ng/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
25kDa
5 x 5μg
TNF (Tumor Necrosis Factor)
TNFSF9, 4-1BB-L, 4-1BBL, AI848817, Cd137l, Ly63l
TNFSF9
Cytokines & Receptors/CD Proteins
His Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Tumor necrosis factor (ligand) superfamily, member 9
Assay Standard
Mammalian
Videos