missing translation for 'onlineSavingsMsg'
Learn More

Thermo Scientific™ Sino Biological™ TNFSF13B Recombinant Human Protein, hIgG1-Fc Tag

This recombinant protein was expressed from a DNA sequence encoding the soluble form of human TNFSF13B (Q9Y275-1) (Ala 134-Leu 285) fused to the Fc region of human IgG1 at the N-terminus

Brand:  Thermo Scientific™ 10056H01H25

Code : NEW

Additional Details : Weight : 0.10000kg

 View more versions of this product

Product Code. 15204565

  • £205.08 / 25µg


Estimated Shipment
14-06-2018
Add to basket

Description

Description

Tumor Necrosis Factor Super Family 13 subunit beta (TNFSF13B), recombinant human/mouse protein is supplied as a lyophilized powder. It is suitable for use in protein studies such as protein structure analysis and protein-protein interactions. In general, recombinant proteins can also be used as an immunogen, as a protein standard, or in cell biology research applications.
  • Activity: Measured by its binding ability in a functional ELISA; immobilized recombinant human TNFSF13B at 1μg/mL (100μL/well) can bind human TNFRSF17, the EC50 of human TNFRSF17 is 0.07μg/mL
  • Formulation: Lyophilized in 140mM NaCl, 2.7mM KCl, 10mM Na2HPO4, 1.8mM KH2PO4, pH 7.4, 5% Mannitol, 5% Trehalose, 0.02% Tween 80
  • Reconstitution: Dissolve the protein in sterile double distilled water to a concentration of 0.2mg/mL or lower; aliquot and use as soon as possible

This protein is manufactured by Sino Biological Inc.

Specifications

Specifications

Proteins, Expression, Isolation & Analysis; Enzyme & Protein Activity Assays; Protein Assay Controls, Reference Standards & Accessories
43.7kDa
5 x 5μg
TNF (Tumor Necrosis Factor)
TNFSF13B, BAFF, BLYS, CD257, DTL, TALL-1, TALL1, THANK, TNFSF20, ZTNF4
TNFSF13B
Cytokines & Receptors/CD Proteins
IgG1 Fc Tag
Lyophilized
≥90% by SDS-PAGE
Store at -20°C. Store aliquots under sterile conditions at -20°C. Avoid repeated freeze-thaw cycles. Expires one year from date of receipt when stored as instructed.
<1 EU/μg
Tumor necrosis factor (ligand) superfamily, member 13b
Assay Standard
Mammalian
Videos

For Research Use Only. Not for use in diagnostic procedures.